missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZIP14 Recombinant Rabbit Monoclonal Antibody (1J6T5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558002
This item is not returnable.
View return policy
Description
Immunogen sequence: SSLGAPAISA ASFLQDLIHR YGEGDSLTLQ QLKALLNHLD VGVGRGNVTQ HVQGHRNLST CFSSGDLFTA HNFSEQSRIG SSELQEFCPT ILQQLDSRAC TSENQENEEN EQTEEGRPSA VEVWGYG.
The zinc transporter ZIP14, also known as SLC39A14, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP14 is a glycosylated multipass plasma membrane protein that belongs to the ZIP transporter subfamily LIV-1. ZIP14 has been shown to contribute to the hypozincemia of inflammation and infection and is regulated in the liver by IL-6. In addition to zinc, ZIP14 is also involved in the cellular uptake of non-transferrin-bound iron as well as iron bound to transferrin.
Specifications
| ZIP14 | |
| Recombinant Monoclonal | |
| 0.52 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q15043, Q75N73 | |
| SLC39A14 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1J6T5 | |
| Unconjugated | |
| SLC39A14 | |
| cig19; Factor for adipocyte differentiation 123; Fad123; FAD-123; HMNDYT2; Kiaa0062; LIV-1 subfamily of ZIP zinc transporter 4; LZT-Hs4; NET34; SLC39A14; solute carrier family 39 (metal ion transporter), member 14; solute carrier family 39 (zinc transporter), member 14; solute carrier family 39 member 14; Zinc transporter ZIP14; Zip14; ZIP-14; Zrt- and Cig19; Zrt- and Irt-like protein 14; Zrt-, Irt-like protein 14 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 213053, 23516, 306009 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction