missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZIP12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12829222
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12829222 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12829222 Supplier Invitrogen™ Supplier No. PA521075

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human spleen tissue lysate. PA5-21075 can be used with blocking peptide PEP-1189.

The zinc transporter ZIP12, also known as SLC39A12, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP12 is a multipass membrane protein that belongs to the ZIP transporter subfamily LIV-1. Increased levels of ZIP12 have been observed during acute inflammation in the airway and asthma, suggesting that it may be promote increased zinc uptake, as this can reduce inflammation. The zinc content in brains in individuals with schizophrenia is lower than in individuals with other cerebral diseases, and mutations in ZIP12 have been observed in a small group of schizophrenic patients, suggesting that ZIP12 may play a role in this disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIP12
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC39A12
Gene Accession No. Q504Y0, Q5FWH7
Gene Alias AW046938; bA570F3.1; Gm731; LIV-1 subfamily of ZIP zinc transporter 8; LZT-Hs8; Slc39a12; solute carrier family 39 (metal ion transporter), member 12; solute carrier family 39 (zinc transporter), member 12; solute carrier family 39 member 12; Zinc transporter ZIP12; Zip12; ZIP-12; Zrt- and Irt-like protein 12
Gene Symbols SLC39A12
Host Species Rabbit
Immunogen A 15 amino acid peptide near the amino terminus of human ZIP12.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221074, 277468
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.