missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZIP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590883
This item is not returnable.
View return policy
Description
Immunogen sequence: MCFRTKLSVS WVPLFLLLSR VFSTETDKPS AQDSRSRGSS GQPADLLQVL SAGDHPPHNH SRSLIKTLLE KTGCPRRRNG MQGDCNLCFE PDALLLIAGG; Positive Samples: 293T, mouse brain, mouse eye, mouse lung, rat brain.
The zinc transporter ZIP12, also known as SLC39A12, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP12 is a multipass membrane protein that belongs to the ZIP transporter subfamily LIV-1. Increased levels of ZIP12 have been observed during acute inflammation in the airway and asthma, suggesting that it may be promote increased zinc uptake, as this can reduce inflammation. The zinc content in brains in individuals with schizophrenia is lower than in individuals with other cerebral diseases, and mutations in ZIP12 have been observed in a small group of schizophrenic patients, suggesting that ZIP12 may play a role in this disease.
Specifications
| ZIP12 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A12 | |
| AW046938; bA570F3.1; Gm731; LIV-1 subfamily of ZIP zinc transporter 8; LZT-Hs8; Slc39a12; solute carrier family 39 (metal ion transporter), member 12; solute carrier family 39 (zinc transporter), member 12; solute carrier family 39 member 12; Zinc transporter ZIP12; Zip12; ZIP-12; Zrt- and Irt-like protein 12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 221074, 277468, 291328 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.07 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q504Y0, Q5FWH7 | |
| SLC39A12 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC39A12 (NP_0011386671). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction