missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZIP11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12858822
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858822 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858822 Supplier Invitrogen™ Supplier No. PA520679

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is mouse kidney tissue lysate. PA5-20679 can be used with blocking peptide PEP-0797.

ZIP11, also known as Slc39A11, is a member of the broader ZIP family of multi-transmembrane domain metal ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. Members of the ZIP family generally transport metal ions from the cell exterior or lumen of intracellular organelles into the cytoplasm, as opposed to the ZnT (SLC30) family of zinc transporters that serve to reduce intracellular zinc availability by promoting zinc efflux from cells or into intracellular vesicles. At least three isoforms of ZIP11 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIP11
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC39A11
Gene Accession No. Q6P6S2, Q8BWY7, Q8N1S5
Gene Alias 1810074D23Rik; C17orf26; SLC39A11; solute carrier family 39 (metal ion transporter), member 11; solute carrier family 39 member 11; solute carrier family 39, member 11; zinc transporter ZIP11; ZIP11; ZIP-11; Zrt- and Irt-like protein 11
Gene Symbols SLC39A11
Host Species Rabbit
Immunogen A 15 amino acid peptide near the center of the human ZIP11.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201266, 287796, 69806
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.