missing translation for 'onlineSavingsMsg'
Learn More

ZIP10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12829212
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12829212 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12829212 Supplier Invitrogen Supplier No. PA521064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ZIP10 Polyclonal Antibody

ZIP10 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIP10
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC39A10
Gene Accession No. Q6P5F6, Q9ULF5
Gene Alias 2900042E17Rik; 5430433I10; Kiaa1265; LZT-Hs2; mKIAA1265; Slc39a10; solute carrier family 39 (metal ion transporter), member 10; solute carrier family 39 (zinc transporter), member 10; solute carrier family 39 member 10; Zinc transporter ZIP10; ZIP10; ZIP-10; Zrt- and Irt-like protein 10
Gene Symbols SLC39A10
Host Species Rabbit
Immunogen An 18 amino acid peptide near the center of human ZIP10.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227059, 363229, 57181
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.