missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIP10 Polyclonal Antibody
ZIP10 Polyclonal Antibody
Brand: Thermo Scientific PA557696
This item is not returnable.
View return policy
Description
ZIP10 Polyclonal Antibody for ICC/IF, IHC
Specifications
| ZIP10 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A10 | |
| 2900042E17Rik; 5430433I10; Kiaa1265; LZT-Hs2; mKIAA1265; Slc39a10; solute carrier family 39 (metal ion transporter), member 10; solute carrier family 39 (zinc transporter), member 10; solute carrier family 39 member 10; Zinc transporter ZIP10; ZIP10; ZIP-10; Zrt- and Irt-like protein 10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57181 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULF5 | |
| SLC39A10 | |
| Recombinant protein corresponding to Human ZIP10. Recombinant protein control fragment (Product #RP-102722). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction