missing translation for 'onlineSavingsMsg'
Learn More

Zincon monosodium salt

Product Code. p-7027476
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11451307

Brand: Thermo Scientific Alfa Aesar B24256.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62625-22-3 | C20H16N4NaO6S | 463.42 g/mol

Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.

Solubility
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-22-3
Molecular Formula C20H16N4NaO6S
Molecular Weight (g/mol) 463.42
MDL Number MFCD00064385
InChI Key IABRINWJUBAIIE-JJECXDOKSA-N
PubChem CID 131856391
IUPAC Name 2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium
SMILES C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na]

Specifications

Melting Point ∽203°C (decomposition)
Quantity 1 g
Beilstein 3872699
Solubility Information Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.
Formula Weight 462.42
Chemical Name or Material Zincon monosodium salt
compliance-icons
Product Identifier
  • Zincon monosodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Zincon monosodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.