Learn More
Bochem™ Zinced Pinchcock with Aluminum Screw
Product Code. 10182501
Change view
Click to view available options
Diameter (Metric):
15 mm
20 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10182501 15 mm Each
10428432 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10182501 Supplier Bochem™ Supplier No. 12121
Only null left
Add to basket
Add to basket
Restrict or regulate tubing fluid flow with these zinced, blade-style pinchcocks. Bochem™ Zinced Pinchcocks facilitates precise flow adjustment.
  • Precisely and easily adjust flow.
  • Durable and resistant.
Specifications
Diameter (Metric) 15 mm
Material Zinc, Aluminum
Product Type Pinchcock