missing translation for 'onlineSavingsMsg'
Learn More

ZINC03129319, MedChemExpress

Product Code. 16483156
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483156 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483156 Supplier Medchem Express Supplier No. HY112254

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress ZINC03129319 is a dengue virus (DENV) NS2B-NS3 protease inhibitor extracted from patent US20150141521A1, has inhibition constants (Ki1) of 92 μM and Ki3 of 20 μM.

ZINC03129319 is a dengue virus (DENV) NS2B-NS3 protease inhibitor extracted from patent US20150141521A1, has inhibition constants (Ki1) of 92 μM and Ki3 of 20 μM.

Specifications

Chemical Name or Material ZINC03129319
CAS 1777807-64-3
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C24H14N2O6S2
Quantity 25 mg
Solubility Information DMSO : 41.67 mg/mL (84.95 mM; Need ultrasonic)
SMILES O=C1C2(SC[C@H]3N2C([C@@H]4N(C5(C(C(C=CC=C6)=C6C5=O)=O)SC4)C3=O)=O)C(C7=CC=CC=C71)=O
Molecular Weight (g/mol) 490.51
Formula Weight 490.51
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.