missing translation for 'onlineSavingsMsg'
Learn More

Zinc wire, 0.5mm dia., 99.994% (metals basis), Puratronic™, Thermo Scientific Chemicals

Catalog Number p-5547536
Click to view available options
Quantity:
1 m
25 m
5 m
This item is not returnable. View return policy
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]
This item is not returnable. View return policy
7440-66-6
Zn
65.38
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
23994
CHEBI:30185
zinc
[Zn]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance Gray wire
Density 7.14g/cm3
Assay Percent Range 99.994% (Metals basis)
Quantity 1 m
Odor Odorless
Form Wire
Chemical Name or Material Zinc

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'