missing translation for 'onlineSavingsMsg'
Learn More

Zinc tungsten oxide, 99.9% (metals basis)

Product Code. 11339397
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339397 50 g 50g
11349397 10 g 10g
11353748 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339397 Supplier Thermo Scientific Alfa Aesar Supplier No. 040234.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13597-56-3 | O4WZn | 313.216 g/mol

It is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13597-56-3
Molecular Formula O4WZn
Molecular Weight (g/mol) 313.216
MDL Number MFCD00049633
InChI Key LJSXSLGFYGBEPA-UHFFFAOYSA-N
Synonym zinc tungsten oxide, zinc tungstate, zinkwolframat, wo4.zn, zinc 2+ tungstate, zinc, ion zn2+ tungstate
PubChem CID 22022519
IUPAC Name zinc;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[Zn+2]

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 313.22
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Zinc tungsten oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.