missing translation for 'onlineSavingsMsg'
Learn More

Zinc trifluoromethanesulfonate, 98%

Product Code. 10414545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10414545 50 g Glass bottle 50g
10618221 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10414545 Supplier Thermo Scientific Acros Supplier No. 317090500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54010-75-2 | C2F6O6S2Zn | 363.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54010-75-2
Molecular Formula C2F6O6S2Zn
Molecular Weight (g/mol) 363.51
MDL Number MFCD00013229
InChI Key CITILBVTAYEWKR-UHFFFAOYSA-L
Synonym zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
PubChem CID 104671
IUPAC Name zinc;trifluoromethanesulfonate
SMILES [Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Color Gray to White
Physical Form Powder
Quantity 50 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula (CF3SO3)2Zn
Solubility Information Solubility in water: very soluble
Formula Weight 363.51
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Zinc trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Zinc trifluoromethanesulphonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Zinc trifluoromethanesulphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.