missing translation for 'onlineSavingsMsg'
Learn More

Zinc titanium oxide, 99.9% (metals basis)

Product Code. 11358818
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358818 250 g 250g
11368818 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358818 Supplier Thermo Scientific Alfa Aesar Supplier No. 041673.30
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12036-69-0 | O4TiZn2 | 242.62 g/mol

Zinc titanium oxide is used as a regenerable catalyst, a pigment and a sorbent of sulfur compounds. It is also used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-69-0
Molecular Formula O4TiZn2
Molecular Weight (g/mol) 242.62
MDL Number MFCD00054107
InChI Key ZBFOLPMOGPIUGP-UHFFFAOYSA-N
Synonym Zinc titanate

Specifications

Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 242.64
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc titanium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.