missing translation for 'onlineSavingsMsg'
Learn More

Zinc telluride, 99.99% (metals basis)

Product Code. 11468610
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468610 25 g 25g
11448610 5 g 5g
11458610 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468610 Supplier Thermo Scientific Alfa Aesar Supplier No. 044412.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1315-11-3 | TeZn | 192.98 g/mol

Zinc telluride can be easily doped, and for this reason it is one of the more common semiconducting materials used in optoelectronics. ZnTe is important for development of various semiconductor devices, including blue LEDs, laser diodes, solar cells, and components of microwave generators. It can be used for solar cells, for example, as a back-surface field layer and p-type semiconductor material for a CdTe/ZnTe structure[6] or in PIN diode structures. The material can also be used as a component of ternary semiconductor compounds.Zinc telluride together with lithium niobate is often used for generation of pulsed terahertz radiation in time-domain terahertz spectroscopy and terahertz imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-11-3
Molecular Formula TeZn
Molecular Weight (g/mol) 192.98
MDL Number MFCD00043079
InChI Key NSRBDSZKIKAZHT-UHFFFAOYSA-N
Synonym zinc telluride, zinc telluride znte, unii-ir8eb6g3vq, zinktellurid, ir8eb6g3vq, zinc telluride,-100 mesh trace metals basis
PubChem CID 3362486
IUPAC Name tellanylidenezinc
SMILES [Zn]=[Te]

Specifications

Melting Point 1240°C
Color Red
Physical Form Powder
Quantity 25 g
Merck Index 14,10162
Solubility Information It decomposes in water.
Formula Weight 192.97
Percent Purity 99.99%
Odor Odorless
Refractive Index 3.56
Density 5.9 g/mL
Assay (metals basis)
Chemical Name or Material Zinc telluride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.