missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Technical, Powder, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
2 kg
Unit Size:
1kg
2kg
500g
Specifications
Specifications
| Color | Gray |
| Melting Point | 420°C |
| Boiling Point | 907°C |
| Quantity | 500 g |
| Packaging | Plastic powder jar |
| Vapor Pressure | 1.3mbar 478 |
| Form | Powder/Solid |
| Formula Weight | 65.36g/mol |
Safety and Handling
Product Identifier
- Zinc, powder
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 1
- Pyrophoric solid Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use sand to extinguish.
- P273-Avoid release to the environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction