Learn More
Zinc sulfide, 99.995% (metals basis)
Description
Zinc sulfide is used in preparation of flexible transparent conductive coatings essential for fabrication of a variety of printed electronic devices such as flexible displays and solar cells. Used to prepare a composite CdS-ZnS/Zirconium-titanium phosphate (ZTP) photocatalyst for hydrogen production under visible light and also applied for light-controlled bioelectrochemical sensors based on CdSe/ZnS quantum dots. It is used for preparation of color tunable light-emitting diodes (LEDs) and also applied to prepare (CdS-ZnS)-TiO2 combined photocatalysts for electricity production via photoelectrocatalysis. It is used as a catalyst for synthesis of spirooxindole derivatives in aqueous medium via Knoevenagel condensation followed by Michael addition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1700°C |
| Physical Form | Pieces |
| Quantity | 250 g |
| Assay Percent Range | 99.995% (metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 97.43 |
| Density | 4.1 |
| Chemical Name or Material | Zinc sulfide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.