Learn More
Zinc sulfide, 99.99% (metals basis), Thermo Scientific Chemicals
Used in Luminescent material, infra red optical material, semiconductor, as a pigment, used as a window for visible optics and infrared optics.
£19.30 - £126.00
Chemical Identifiers
CAS | 1314-98-3 |
---|---|
Molecular Formula | SZn |
Molecular Weight (g/mol) | 97.44 |
MDL Number | MFCD00011301 |
InChI Key | DRDVZXDWVBGGMH-UHFFFAOYSA-N |
Synonym | zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide |
PubChem CID | 14821 |
SMILES | [S--].[Zn++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379227
|
Thermo Scientific Alfa Aesar
040091.09 |
10 g |
£19.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369227
|
Thermo Scientific Alfa Aesar
040091.18 |
50 g |
£41.10
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359227
|
Thermo Scientific Alfa Aesar
040091.30 |
250 g |
£126.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in Luminescent material, infra red optical material, semiconductor, as a pigment, used as a window for visible optics and infrared optics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-98-3 | |
97.44 | |
DRDVZXDWVBGGMH-UHFFFAOYSA-N | |
14821 |
SZn | |
MFCD00011301 | |
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide | |
[S--].[Zn++] |
Specifications
1700°C | |
White | |
99.99% metal basis | |
MFCD00011301 | |
soluble in acids,insoluble in water. | |
[S--].[Zn++] | |
97.44 | |
97.43 | |
4.1 |
1314-98-3 | |
Powder | |
SZn | |
zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide | |
DRDVZXDWVBGGMH-UHFFFAOYSA-N | |
sulfanylidenezinc | |
14821 | |
Odorless | |
Zinc sulfide |
Safety and Handling
missing translation for 'einecsNumber' : 215-251-3
missing translation for 'rtecsNumber' : ZH5400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only