missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zinc sulfide, 99.99% (metals basis)

Brand:  Alfa Aesar™ 013239.30

Code : NEW

Additional Details : CAS Number : 1314-98-3 Weight : 0.25000kg

Quantity 250g

 View more versions of this product

Product Code. 11326688

  • £246.10 / 250g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Zinc sulfide
ZnS
MFCD00011301
1314-98-3
97.43
14,10160

Specifications

Zinc sulfide
Lumps
(metals basis)
97.43
14,10160
Odorless
White
250g
1314-98-3
99.99%
ZnS
MFCD00011301
4.10g/mL
2 to 20mm
1700°C
Videos
SDS