missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11316688
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Ziinc sulfide is used in luminescent material, infra red optical material, semiconductor, as a pigment, used as a window for visible optics and infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-98-3
Molecular Formula SZn
Molecular Weight (g/mol) 97.44
MDL Number MFCD00011301
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
PubChem CID 14821
IUPAC Name sulfanylidenezinc
SMILES [S--].[Zn++]

Specifications

Melting Point 1700°C
Color White
Physical Form Lump
Quantity 50 g
Linear Formula ZnS
Merck Index 14,10160
Solubility Information Soluble in acids. insoluble in water.
Formula Weight 97.43
Percent Purity 99.99%
Odor Odorless
Density 4.10 g/mL
Assay (metals basis)
Chemical Name or Material Zinc sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'