Learn More
Zinc sulfate monohydrate, Zn 35.5%
Descrizione
Zinc sulfate monohydrate is used in calico-printing, preserving wood and skins, electrolytes for zinc plating, bleaching paper and clarifying glue. It is also used as a chemical reagent, coagulant in the preparation of rayon, mordant in dyeing and as a zinc source in animal feeds. In medicine, it is used as an astringent and emetic. Further, it is a precursor to the pigment lithopone. In addition to this, it is used to supply zinc in fertilizers, agricultural sprays, in electrolytes for zinc plating and as a mordant in dyeing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Physical Form | Coarse Powder |
| Quantity | 1 kg |
| UN Number | UN3077 |
| Merck Index | 14,10159 |
| Solubility Information | Soluble in water and alcohol. |
| Formula Weight | 179.45 (161.43 Anhydrous) |
| Concentration or Composition (by Analyte or Components) | Zn 35.5% |
| Chemical Name or Material | Zinc sulfate monohydrate |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.