missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate Monohydrate, purum p.a., ≥99% (KT), Honeywell™
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| Melting Point | 238°C |
| Color | Colorless |
| Physical Form | Solid |
| Quantity | 250 g |
| Linear Formula | ZnSO4 · H2O |
| UN Number | UN3077 |
| Solubility Information | In water 538g/L at 20°C |
| Formula Weight | 179.47g/mol |
| Percent Purity | ≥99% (KT) |
| Odor | Odorless |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?