missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate monohydrate, 99%

Product Code. 10136813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10136813 5 kg Plastic bottle 5kg
10559993 250 g Plastic bottle 250g
10656124 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10136813 Supplier Thermo Scientific Acros Supplier No. 389800050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7446-19-7 | O4SZn·H2O | 179.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-19-7
Molecular Formula O4SZn·H2O
Molecular Weight (g/mol) 179.47
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
IUPAC Name zinc;sulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[Zn+2]

Specifications

Color White
Physical Form Powder
Quantity 5 kg
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula ZnSO4·H2O
Merck Index 15, 10359
Solubility Information Solubility in water: 350g/L (20°C). Other solubilities: practically insoluble in ethanol
Formula Weight 179.47
Percent Purity 99%
Identification Confirmed (Zinc), Confirmed (Sulfate)
Packaging Plastic bottle
Chemical Name or Material Zinc sulfate monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.