missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Monohydrate, ≥99.9% trace metals basis, Honeywell™

Product Code. 15693120 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15693120 500 g Poly Bottle 500g
15683120 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15693120 Supplier Honeywell Research Chemicals Supplier No. 307491500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

≥99.9% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-19-7
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.451
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
IUPAC Name zinc;sulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[Zn+2]

Specifications

Melting Point 238°C
Quantity 500 g
Linear Formula ZnSO4 · H2O
UN Number UN3077
Solubility Information Soluble in water
Formula Weight 179.47g/mol
Percent Purity ≥99.9% (trace metals basis)
Packaging Poly Bottle
Chemical Name or Material Zinc Sulfate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.