missing translation for 'onlineSavingsMsg'
Learn More

Melford Zinc Sulfate Monohydrate

Product Code. 30018975
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018975 5 kg 5kg
30018974 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018975 Supplier Melford Supplier No. Z223505000.0
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

CAS Number: 7446-19-7

Molecular Formula: ZnSO4 H2O

Molecular Weight: 179.5

Specifications

Chemical Name or Material Zinc Sulfate Monohydrate, USP
CAS 7446-19-7
Quantity 5 kg
Molecular Formula ZnSO4-H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.