missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate hydrate, Puratronic, 99.998% (metals basis)

Product Code. 11366158
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366158 10 g 10g
11376158 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366158 Supplier Thermo Scientific Alfa Aesar Supplier No. 011138.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16788-42-4 | H2O5SZn | 179.45 g/mol

Zinc sulfate hydrate is used in the production of rayon, as a feed supplement, and as a fertilizer ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16788-42-4
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.45
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
SMILES O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 100°C
Color White
Physical Form Crystalline Powder
Quantity 10 g
Linear Formula ZnSO4·xH2O
UN Number UN3077
Merck Index 14,10159
Solubility Information Soluble in water.
IUPAC Name zinc;sulfate;hydrate
Formula Weight 161.43 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc sulfate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.