missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate heptahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Solstice

Product Code. 15693490 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 500 g
25 kg
6 x 1 kg
Unit Size:
1kg
25kg
500g
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15693490 25 kg 25kg
15603500 500 g 500g
15683490 1 kg 1kg
15623500 6 x 500 g Pack of 6
15613500 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15693490 Supplier Honeywell-Fluka Supplier No. 3166525KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 25 kg
Linear Formula ZnSO4 · 7H2O
UN Number 3077
Formula Weight 287.56g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Chemical Name or Material Zinc Sulfate Heptahydrate, Reag. Ph. Eur. reag. ISO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.