missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Heptahydrate, 99.0 to 103.0%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15632120 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15632120 100 g Poly Bottle 100g
15652120 500 g Poly Bottle 500g
15642120 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15632120 Supplier Honeywell-Fluka Supplier No. 221376100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 100 g
Linear Formula ZnSO4 · 7H2O
UN Number UN3077
Solubility Information Soluble in water, 965g/L at 20°C
IUPAC Name zinc;sulfate;heptahydrate
Formula Weight 287.56g/mol
Percent Purity 99.0 to 103.0%
Grade ACS Reagent
Packaging Poly Bottle
Chemical Name or Material Zinc Sulfate Heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.