missing translation for 'onlineSavingsMsg'
Learn More

Zinc Stearate, MP Biomedicals

Product Code. 11403855 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403855 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11403855 Supplier MP Biomedicals Supplier No. 0522003080
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 557-05-1
Molecular Formula C36H70O4Zn
Quantity 100 g
Synonym zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2]
IUPAC Name zinc;octadecanoate
Molecular Weight (g/mol) 632.332
PubChem CID 11178
Formula Weight 632.33
Chemical Name or Material Zinc Stearate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.