missing translation for 'onlineSavingsMsg'
Learn More

Zinc stearate

Product Code. 10202512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
20 kg
Unit Size:
1kg
20kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10202512 5 kg 5kg
10018511 1 kg 1kg
10731804 20 kg 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10202512 Supplier Thermo Scientific Acros Supplier No. 263780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 557-05-1 | C36H70O4Zn | 632.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-05-1
MDL Number MFCD00013031
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
Synonym zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat
PubChem CID 11178
IUPAC Name zinc;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2]

Specifications

Melting Point 118.0°C to 128.0°C
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Packaging Fiber Drum
Molecular Formula C36H70O4Zn
Linear Formula [CH3(CH2)16CO2]2Zn
Quantity 5 kg
Merck Index 15, 10358
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol,diethyl ether,,soluble in benzene,xylene,fats,oils
Molecular Weight (g/mol) 632.34
Formula Weight 632.34
Percent Purity 12.5 to 14.0% (ZnO)
Physical Form Powder
Chemical Name or Material Zinc stearate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.