missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£51.65 - £110.00
Chemical Identifiers
CAS | 7440-66-6, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | Zn |
Molecular Weight (g/mol) | 65.38 |
MDL Number | MFCD00011291 |
InChI Key | HCHKCACWOHOZIP-UHFFFAOYSA-N |
PubChem CID | 23994 |
ChEBI | CHEBI:30185 |
IUPAC Name | zinc |
SMILES | [Zn] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
16301186
|
SPEX Certiprep
CLZN2-2M |
30 mL | Triple-leached LDPE bottle |
£51.65
30mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15628285
|
SPEX Certiprep
CLZN2-2Y |
125 mL | Triple-leached LDPE bottle |
£110.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11496008
|
SPEX Certiprep
CLZN2-2Y |
125 mL | Triple-leached LDPE bottle | ||||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Zn | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-66-6, 7697-37-2, 7732-18-5 | |
65.38 | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
CHEBI:30185 | |
[Zn] |
Zn | |
MFCD00011291 | |
23994 | |
zinc |
Specifications
7440-66-6, 7697-37-2, 7732-18-5 | |
0.1, 2.0, 97.9 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011291 | |
[Zn] | |
65.38 | |
CHEBI:30185 | |
2% HNO3 |
0.1, 2.0, 97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Zn | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
zinc | |
23994 | |
Trace ICP/ICP-MS | |
Zinc Standard |
Safety and Handling
Causes skin irritation, serious eye irritation
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1000 µg/mL Zinc
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Product intended for laboratory use only