missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc standard solution, BAKER INSTRA-ANALYZED reagent for AAS, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
:
100 mL
Specifications
Specifications
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | 1000 μg/mL |
| Packaging | Plastic bottle |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362-Take off contaminated clothing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction