Learn More
Zinc Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£80.60 - £172.00
Chemical Identifiers
CAS | 7440-66-6, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | Zn |
Molecular Weight (g/mol) | 65.38 |
MDL Number | MFCD00011291 |
InChI Key | HCHKCACWOHOZIP-UHFFFAOYSA-N |
PubChem CID | 23994 |
ChEBI | CHEBI:30185 |
IUPAC Name | zinc |
SMILES | [Zn] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10326272
|
SPEX Certiprep
PLZN2-2Y |
125 mL | Triple-leached LDPE bottle |
£80.60
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10366222
|
SPEX Certiprep
PLZN2-2X |
500 mL | Triple-leached LDPE bottle | ||||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Zn | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-66-6, 7697-37-2, 7732-18-5 | |
65.38 | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
CHEBI:30185 | |
[Zn] |
Zn | |
MFCD00011291 | |
23994 | |
zinc |
Specifications
7440-66-6, 7697-37-2, 7732-18-5 | |
1.0, 5.0, 94.0 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Zn | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
zinc | |
23994 | |
Plasma | |
2% HNO3 |
1.0, 5.0, 94.0 | |
Assurance Certificate of Analysis reports up 30 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011291 | |
[Zn] | |
65.38 | |
CHEBI:30185 | |
Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLZN2-2Y, -2T, -2X 10,000 PLZN2-3Y, -3X 2-5% HCl 1000μg/mL PLZN1-2X 10,000 PLZN1-3X |
|
Zinc Standard |
Safety and Handling
Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only