missing translation for 'onlineSavingsMsg'
Learn More

Zinc solution for ICP 1g/L HCl matrix, Fisher chemical™
BIOPHARMA

Product Code. p-7084032 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
This item is not returnable. View return policy

Product Code. 12946164

Brand: Fisher Chemical S 069 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Analyte or Component Names Zn
Concentration 1±0.002 g/L
Type Mass Spectrometry Standard
Element Zn
Solvent or Matrix Hydrogen Chloride
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.