missing translation for 'onlineSavingsMsg'
Learn More

Zinc shot, 1-5mm (0.04-0.2in), 99.999% (metals basis)

Product Code. 11336857
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11336857

Brand: Thermo Scientific Alfa Aesar 010759.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray shot
Quantity 1 kg
Odor Odorless
Assay Percent Range 99.999% (Metals basis)
Form Shot
Formula Weight 65.3
Chemical Name or Material Zinc, 1-5mm (0.04-0.2 in.)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.