missing translation for 'onlineSavingsMsg'
Learn More

Zinc selenite

Product Code. 11396449
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
Unit Size:
100g
10g
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396449 500 g 500g
11316459 10 g 10g
11386449 100 g 100g
11306459 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11396449 Supplier Thermo Scientific Alfa Aesar Supplier No. 012586.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13597-46-1 | O3SeZn | 192.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13597-46-1
Molecular Formula O3SeZn
Molecular Weight (g/mol) 192.35
MDL Number MFCD00011297
InChI Key PEUPCBAALXHYHP-UHFFFAOYSA-L
Synonym zinc selenite, selenious acid zinc salt 1:1, selenious acid, zinc salt 1:1, zinkselenit, acmc-1bpna, zinc 2+ ion selenite, selenious acid, zinc salt, zinc 2+ selenite, zinc, ion zn2+ selenit
PubChem CID 159686
IUPAC Name zinc;selenite
SMILES [Zn++].[O-][Se]([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
UN Number UN2630
Formula Weight 192.33
Chemical Name or Material Zinc selenite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.