missing translation for 'onlineSavingsMsg'
Learn More

Zinc selenide

Product Code. 11458807
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458807 250 g 250g
11438807 10 g 10g
11448807 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458807 Supplier Thermo Scientific Alfa Aesar Supplier No. A19947.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1315-09-9 | SeZn | 144.35 g/mol

Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-09-9
Molecular Formula SeZn
Molecular Weight (g/mol) 144.35
MDL Number MFCD00011296
InChI Key GTLQJUQHDTWYJC-UHFFFAOYSA-N
Synonym zinc selenide znse, znse, zinc selenide, spectro, zinc selenide, powder, 10 mum trace metals basis, zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick
IUPAC Name zinc(2+) selandiide
SMILES [Zn++].[Se--]

Specifications

Melting Point >1100°C
Quantity 250 g
UN Number UN3283
Merck Index 14,10156
Solubility Information Insoluble in water.
Formula Weight 144.33
Odor Odorless
Density 5.42^1^5
Chemical Name or Material Zinc selenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.