Learn More
Zinc selenide
Description
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.
Specifications
Specifications
| Melting Point | >1100°C |
| Quantity | 10 g |
| UN Number | UN3283 |
| Merck Index | 14,10156 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 144.33 |
| Odor | Odorless |
| Density | 5.42^1^5 |
| Chemical Name or Material | Zinc selenide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.