missing translation for 'onlineSavingsMsg'
Learn More

Zinc selenide, Optical Grade, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11369239
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11369239

Brand: Thermo Scientific Alfa Aesar 088298.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-09-9
Molecular Formula SeZn
Molecular Weight (g/mol) 144.35
MDL Number MFCD00011296
InChI Key GTLQJUQHDTWYJC-UHFFFAOYSA-N
Synonym zinc selenide znse, znse, zinc selenide, spectro, zinc selenide, powder, 10 mum trace metals basis, zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick
IUPAC Name zinc(2+) selandiide
SMILES [Zn++].[Se--]

Specifications

Melting Point >1100°C
Physical Form Pieces
Quantity 25 g
UN Number UN3283
Merck Index 14,10156
Formula Weight 144.33
Percent Purity 99.999%
Grade Optical
Odor Odorless
Sensitivity Air Sensitive
Density 5.42 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Zinc selenide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.