missing translation for 'onlineSavingsMsg'
Learn More

Zinc protoporphyrin, 96%

Product Code. 11420530
Change view
Click to view available options
Quantity:
10 mg
50 mg
0.25 g
Unit Size:
0.25g
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420530 0.25 g 0.25g
11358317 10 mg 10mg
11368317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420530 Supplier Thermo Scientific Alfa Aesar Supplier No. 032068.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15442-64-5 | C34H32N4O4Zn | 626.034 g/mol

Zinc protoporphyrin-9 is a potent, selective heme oxygenase inhibitor, interleukin-1 antagonist, GCS inhibitor, and NOS1, NOS2, and NOS3 inhibitor. In olfactory neuronal cultures, this product has been used to block CO biosynthesis, which lowered cGMP concentrations. Zinc protoporphyrin-9 is an extremely useful tool to elucidate the role of CO in signal transduction and as a neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 15442-64-5
Molecular Formula C34H32N4O4Zn
Molecular Weight (g/mol) 626.034
MDL Number MFCD00011612
InChI Key FUTVBRXUIKZACV-UHFFFAOYSA-L
Synonym zinc protoporphyrin
PubChem CID 131664383
IUPAC Name zinc;3-[18-(2-carboxyethyl)-8,13-bis(ethenyl)-3,7,12,17-tetramethyl-23H-porphyrin-21-id-2-yl]propanoate
SMILES CC1=C(C2=CC3=NC(=CC4=C(C(=C(N4)C=C5C(=C(C(=N5)C=C1[N-]2)C)C=C)C)C=C)C(=C3CCC(=O)O)C)CCC(=O)[O-].[Zn+2]

Especificaciones

Color Purple to Red
Odor Odorless
Quantity 0.25 g
Solubility Information Insoluble in water. Soluble in ethanol,and DMSO,pyridine,or dimethyl formamide.
Formula Weight 626.04
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material Zinc protoporphyrin

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.