missing translation for 'onlineSavingsMsg'
Learn More

Zinc phthalocyanine, 95%

Product Code. 11378358
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378358 50 g 50g
11398358 2 g 2g
11388358 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378358 Supplier Thermo Scientific Alfa Aesar Supplier No. 039553.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14320-04-8 | C32H16N8Zn | 577.916 g/mol

Zinc phthalocyanine finds general applications in industry as catalysts, photoconductors and in biomedical field for photodynamic therapy, PDT. ZnPc loaded PLGA nanoparticles is a promising drug delivery system for photodynamic therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14320-04-8
Molecular Formula C32H16N8Zn
Molecular Weight (g/mol) 577.916
MDL Number MFCD00041955
InChI Key PODBBOVVOGJETB-UHFFFAOYSA-N
Synonym zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 %
PubChem CID 2735172
ChEBI CHEBI:51218
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]

Specifications

Color Purple
Odor Odorless
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 577.91
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Zinc phthalocyanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.