missing translation for 'onlineSavingsMsg'
Learn More

Zinc phosphate hydrate, tech.

Product Code. 11307327
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307327 500 g 500g
11317327 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307327 Supplier Thermo Scientific Alfa Aesar Supplier No. 011589.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34807-26-6 | O8P2Zn3 | 386.08 g/mol

Zinc phosphate hydrate is used in dentistry. It is commonly used for luting permanent metal restorations and as a base for dental restorations. Zinc phosphate cement is also used for cementation of inlays, crowns, bridges, and orthodontic appliances and occasionally as a temporary restoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34807-26-6
Molecular Formula O8P2Zn3
Molecular Weight (g/mol) 386.08
MDL Number MFCD00036282
InChI Key LRXTYHSAJDENHV-UHFFFAOYSA-H
Synonym zinc phosphate, delaphos, trizinc diphosphate, zinc orthophosphate, trizinc phosphate, delaphos 2m, bonderite 40, granodine 80, microphos 90, granodine 16nc
PubChem CID 24519

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in acetic acid,dilute mineral acids,ammonia. Insoluble in water
SMILES [Zn++].[Zn++].[Zn++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
IUPAC Name trizinc(2+) diphosphate
Formula Weight 386.05 (Anhydrous)
Grade Technical
Chemical Name or Material Zinc phosphate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.