missing translation for 'onlineSavingsMsg'
Learn More

Zinc peroxide, approximately 50% ZnO{2}

Product Code. 15441547
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441547 500 g 500g
15431547 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441547 Supplier Thermo Scientific Alfa Aesar Supplier No. 039635.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Curing agent, rubber accelerator | CAS: 1314-22-3 | O2Zn | 97.38 g/mol

Zinc peroxide is used as curing agent, rubber accelerator. has been applied in a variety of settings, from medicine to aesthetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-22-3
Molecular Formula O2Zn
Molecular Weight (g/mol) 97.38
MDL Number MFCD00137469
InChI Key IPTOGCUGCFHDSS-UHFFFAOYSA-N
Synonym zinc peroxide, zinc superoxide, zinc dioxide, zinc peroxide un1516 oxidizer, zinc peroxide, medicinal, unii-0i969dvm77, zinc peroxide, medicinal usp, zpo
PubChem CID 14809
SMILES [Zn++].[O-][O-]

Specifications

Melting Point >150°C (decomposition)
Physical Form Powder
Quantity 500 g
UN Number UN1516
Merck Index 14,10149
Solubility Information Insoluble in water.
IUPAC Name hydrogen peroxide;zinc
Formula Weight 97.38
Concentration or Composition (by Analyte or Components) ≈50% ZnO2
Odor Odorless
Chemical Name or Material Zinc peroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.