missing translation for 'onlineSavingsMsg'
Learn More

Zinc perchlorate hexahydrate, Reagent Grade

Product Code. p-7048834
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11317367

Brand: Thermo Scientific Alfa Aesar 011614.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-64-6 | Cl2H12O14Zn | 372.362 g/mol

Zinc perchlorate hexahydrate, efficiently catalyses the esterification between nearly equimolar amounts of carboxylic acids and alcoholsZinc perchlorate hexahydrate [Zn (ClO4) 2· 6H2O] was found to be a highly efficient catalyst for acylation of electron deficient phenols, sterically hindered alcohols and electron deficient and sterically hindered amines. Zinc (II) perchlorate hexahydrate has been found to be a new and efficient catalyst for conjugate addition of thiols to α, β-unsaturated ketones under solvent-free conditions at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-64-6
Molecular Formula Cl2H12O14Zn
Molecular Weight (g/mol) 372.362
MDL Number MFCD00150370
InChI Key PADPILQDYPIHQQ-UHFFFAOYSA-L
Synonym zinc perchlorate hexahydrate, perchloric acid, zinc salt, hexahydrate, 2clo4.zn.6h2o, zinc diperchlorate hexahydrate, zinc perchlorate 6h2o, zincperchloratehexahydrate, zinc 2+ hexahydrate diperchlorate, zinc perchlorate hexahydrate, reagent grade, zinc, ion zn2+ hexahydrate diperchlorate ion, zinc perchlorate hexahydrate trace metals basis 10g
PubChem CID 202253
IUPAC Name zinc;diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Zn+2]

Specifications

Melting Point 106°C
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula Zn(ClO4)2·6H2O
UN Number UN1481
Merck Index 14,10148
Solubility Information Soluble in water, alcohol
Formula Weight 372.37 (264.28 Anhydrous)
Grade Reagent
Odor Odorless
Refractive Index 1.5
Sensitivity Hygroscopic
Density 2.25 g/mL
Chemical Name or Material Zinc perchlorate hexahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Zinc perchlorate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Zinc perchlorate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.