missing translation for 'onlineSavingsMsg'
Learn More

Zinc perchlorate hexahydrate, 99.997% (metals basis)

Product Code. 11488410
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488410 50 g 50g
11498410 10 g 10g
11408420 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488410 Supplier Thermo Scientific Alfa Aesar Supplier No. 044315.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-64-6 | Cl2H12O14Zn | 372.362 g/mol

Used as intermediates. A synthetic reagent for proteomics research. Zinc(II) perchlorate hexahydrate is a highly efficient catalyst for the opening of epoxides by amines affording 2-amino alcohols in high yields under solvent-free conditions and with excellent chemo-, regio-, and stereoselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-64-6
Molecular Formula Cl2H12O14Zn
Molecular Weight (g/mol) 372.362
MDL Number MFCD00150370
InChI Key PADPILQDYPIHQQ-UHFFFAOYSA-L
Synonym zinc perchlorate hexahydrate, perchloric acid, zinc salt, hexahydrate, 2clo4.zn.6h2o, zinc diperchlorate hexahydrate, zinc perchlorate 6h2o, zincperchloratehexahydrate, zinc 2+ hexahydrate diperchlorate, zinc perchlorate hexahydrate, reagent grade, zinc, ion zn2+ hexahydrate diperchlorate ion, zinc perchlorate hexahydrate trace metals basis 10g
PubChem CID 202253
IUPAC Name zinc;diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Zn+2]

Specifications

Melting Point 106°C
Color White
Physical Form Crystalline
Quantity 50 g
Linear Formula Zn(ClO4)2·6H2O
UN Number UN1481
Merck Index 14,10148
Solubility Information Soluble in ethyl alcohol and water.
Formula Weight 372.37 (264.28 Anhydrous)
Percent Purity 99.997%
Odor Odorless
Refractive Index 1.5
Density 2.25 g/mL
Assay (metals basis)
Chemical Name or Material Zinc perchlorate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.