missing translation for 'onlineSavingsMsg'
Learn More

Zinc p-toluenesulfonate hydrate, Thermo Scientific™

Product Code. 10283052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10283052 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10283052 Supplier Thermo Scientific Acros Supplier No. 371080250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 123334-05-4
Packaging Glass bottle
Molecular Formula C14H14O6S2Zn
Linear Formula (CH3C6H4SO3)2Zn·H2O
MDL Number MFCD00151013
Quantity 25 g
Synonym zinc p-toluenesulfonate hydrate, acmc-20eh0l, p-toluenesulfonic acid zinc salt, zinc 2+ hydrate ditosylate, p-toluenesulfonic acid zinc salt hydrate, benzenesulfonic acid,4-methyl-, zinc salt, hydrate 2:1:?
Solubility Information Solubility in water: insoluble
InChI Key YISPIDBWTUCKKH-UHFFFAOYSA-L
SMILES [Zn++].CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O
IUPAC Name zinc(2+) bis(4-methylbenzene-1-sulfonate)
Molecular Weight (g/mol) 407.76
PubChem CID 16212267
Formula Weight 407.78
Chemical Name or Material Zinc p-toluenesulfonate hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.