missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Oxide, puriss. p.a., ACS Reagent, ≥99% (KT), Honeywell Fluka™
Puriss. p.a., ACS Reagent,≥99.0% (KT)
£25.78 - £75.91
Chemical Identifiers
CAS | 1314-13-2 |
---|---|
Molecular Formula | OZn |
Molecular Weight (g/mol) | 81.38 |
MDL Number | MFCD00011300 |
InChI Key | XLOMVQKBTHCTTD-UHFFFAOYSA-N |
Synonym | zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 |
PubChem CID | 14806 |
ChEBI | CHEBI:36560 |
IUPAC Name | oxozinc |
SMILES | O=[Zn] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15627580
|
Honeywell-Fluka
96479-100G |
100 g | Poly Bottle | ||||||
15647580
|
Honeywell-Fluka
96479-250G |
250 g | Poly Bottle | ||||||
15657580
|
Honeywell-Fluka
96479-500G |
500 g | Poly Bottle |
£55.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15637580
|
Honeywell-Fluka
96479-1KG |
1 kg | Poly Bottle | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1314-13-2 | |
81.38 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
14806 | |
oxozinc |
OZn | |
MFCD00011300 | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
CHEBI:36560 | |
O=[Zn] |
Specifications
∼1975°C | |
White | |
OZn | |
MFCD00011300 | |
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
O=[Zn] | |
81.38 | |
CHEBI:36560 | |
≥99% (KT) | |
5.6g/cm3 at 20°C |
1314-13-2 | |
∼7 (at 20°C) | |
ZnO | |
UN3077 | |
XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
oxozinc | |
14806 | |
81.39g/mol | |
Powder | |
Zinc oxide |
Safety and Handling
P273-P501
missing translation for 'dotInformation' : 9/PGIII
missing translation for 'rtecsNumber' : 215-222-5