missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Oxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
2kg
500g
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 2 kg |
| Formula Weight | 81.39 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 5.61kg |
| Chemical Name or Material | Zinc Oxide |
Safety and Handling
Signal Word
- Warning
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction