missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc oxide, NanoArc™ ZN-0625, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Zinc oxide dispersion in Dowanol PMA suitable for incorporation in a wide variety of coatings formulations
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Zinc oxide dispersion in Dowanol™ PMA suitable for incorporation in a wide variety of coatings formulations
Specifications
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 50% in 1, 2-propanediol monomethyl ether acetate |
UN Number | UN1993 |
Formula Weight | 81.37 |
Odor | Ester-like |
Chemical Name or Material | Zinc oxide, NanoTek™ Z1102PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'