missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, NanoTek∣r

Product Code. 11440880
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440880 100 g 100g
11450880 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440880 Supplier Thermo Scientific Alfa Aesar Supplier No. 044899.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-13-2 | OZn | 81.38 g/mol

NanoTek™ zinc oxide, due to the small particle size, can provide UV protection while maintaining a high level of transparency in coatings, polymers, caulks, adhesives and other resin systemsZinc oxide nanoparticles are used in the manufacture of rubber and cigarettes. It plays an important role in paints as a coating agent. It is used in creams and ointments, which are used to treat skin diseases. It acts as an additive in the manufacture of concrete. It is also used in ceramic industry and as an additive in food products such as breakfast cereals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975°C
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water. Soluble in dilute acetic acid,mineral acids,ammonia,ammonium carbonate and fixed alkali hydroxide solution.
Formula Weight 81.37
Density 5.6
Chemical Name or Material Zinc Oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.