missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide based sulfur removal material, HiFUEL∣r A310

Product Code. 15436945
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436945 2.5 kg 2.5kg
15416945 500 g 500g
15426945 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15436945 Supplier Thermo Scientific Alfa Aesar Supplier No. 045469.A4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Zinc oxide is used in laser diodes and light emitting diodes. It helps in removal of sulfur and sulfur compounds from a variety of fluids and gases particularly industrial flue gases.It acts as a precursor to other zinc compounds and additive to protect the rubber from fungi and UV light. It is mixed with stearic acid and used in the vulcanization of rubber. It is used in products like baby powder, barrier creams to treat diaper rashes, calamine cream, anti-dandruff shampoos, and antiseptic ointments. It is used in nuclear pressurized water reactors to prevent corrosion. It acts as a food additive in cereals and prepackaged foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc oxide based sulfur removal material HiFUEL™ A310
MDL Number MFCD00011300
Quantity 2.5 kg
Beilstein EM>6,
Solubility Information Soluble in hydrochloric acid. Insoluble in water.
Physical Form Spheres

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.